Learn More
Novus Biologicals™ Recombinant Human DNA Polymerase iota GST (N-Term) Protein
Artikelnummer. 18640181 Alle ansehen Bio Techne Produkte
Click to view available options
Menge:
10 μg
25 μg
Packungsgröße:
10 Mikrogramm
25 Mikrogramm
Artikelnummer. 18640181
Marke: Novus Biologicals™ H00011201P0110ug
om dit product te kopen Registreer vandaag om een webaccount aan te maken
Recombinant Protein
This protein was produced in an in vitro wheat germ expression system that should preserve correct conformational folding that is necessary for biological function. While it is possible that this protein could display some level of activity, the functionality of this protein has not been explicitly measured or validated.
TRUSTED_SUSTAINABILITY
Spezifikation
Zur Verwendung mit (Anwendung) Western Blot, ELISA, Protein Array, Immunoaffinity Purification
Zusammensetzung 50 mM Tris-HCl, 10 mM reduced Glutathione, pH 8.0 in the elution buffer.
Gen-ID (Entrez) 11201
Molekulargewicht MolecularWeight-theroretical: 106.7 kDa
Menge 10 μg
Lagerungsbedingungen Store at -80°C. Avoid freeze-thaw cycles.
Gen-Alias DNA polymerase iota, EC 2.7.7.7, Eta2, polymerase (DNA directed) iota, RAD30 homolog B, RAD30Bpolymerase (DNA-directed), iota, RAD3OB
Gensymbol POLI
Forschungskategorie DNA Polymerases, DNA Repair
Reinheits- oder Qualitätsgrad >80% by SDS-PAGE and Coomassie blue staining
Protein DNA Polymerase iota