missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human Cyclin B1 Protein
A DNA sequence encoding the Homo sapiens (Human) Cyclin B1, was expressed in the hosts and tags indicated.
Marke: enQuireBio™ QP10033-ba-100ug
Additional Details : Gewicht : 0.01000kg
Spezifikation
NP_114172.1 | |
Cyclin B1 Protein | |
Research Use Only | |
CCNB1 | |
Recombinant Protein | |
Baculovirus | |
MALRVTRNSK INAENKAKIN MAGAKRVPTA PAATSKPGLR PRTALGDIGN KVSEQLQAKM PMKKEAKPSA TGKVIDKKLP KPLEKVPMLV PVPVSEPVPE PEPEPEPEPV KEEKLSPEPI LVDTASPSPM ETSGCAPAEE DLCQAFSDVI LAVNDVDAED GADPNLCSEY VKDIYAYLRQ LEEEQAVRPK YLLGREVTGN MRAILIDWLV QVQMKFRLLQ ETMYMTVSII DRFMQNNCVP KKMLQLVGVT AMFIASKYEE MYPPEIGDFA FVTDNTYTKH QIRQMEMKIL RALNFGLGRP LPLHFLRRAS KIGEVDVEQH TLAKYLMELT MLDYDMVHFP PSQIAAGAFC LALKILDNGE WTPTLQHYLS YTEESLLPVM QHLAKNVVMV NQGLTKHMTV KNKYATSKHA KISTLPQLNS ALVQDLAKAV AKV | |
Greater than 80% as determined by SDS-PAGE. |
891 | |
100 μg | |
Not determined. | |
Full Length | |
Human | |
His | |
Tris-based buffer, 50% glycerol |