missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human CTF1 Protein
A cDNA sequence encoding the CTF1 was constructed and used to recombinantly synthesize the protein.
Marke: enQuireBio™ QP10564-1mg
Additional Details : Gewicht : 0.01000kg
Spezifikation
1489 | |
1 mg | |
< 1.0 EU per ug protein as determined by the LAL method. | |
The ED50 as determined by a cell proliferation assay using human TF-1 cells is less than 1.0 ng/ml, corresponding to a specific activity of > 1.0M 106 IU/mg. | |
Human | |
Untagged | |
CTF-1 protein was lyophilized from a 0.2microm filtered concentrated solution in 30% Acetonitrile and 0.1% TFA. |
CTF1 Protein | |
Research Use Only | |
CTF1 | |
Recombinant Protein | |
E. coli | |
MSRREGSLED PQTDSSVSLL PHLEAKIRQT HSLAHLLTKY AEQLLQEYVQ LQGDPFGLPS FSPPRLPVAG LSAPAPSHAG LPVHERLRLD AAALAALPPL LDAVCRRQAE LNPRAPRLLR RLEDAARQAR ALGAAVEALL AALGAANRGP RAEPPAATAS AASATGVFPA KVLGLRVCGL YREWLSRTEG DLGQLLPGGS A | |
Greater than 95.0% as determined by:(a) Analysis by RP-HPLC. (b) Analysis by SDS-PAGE. |