missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human CKMBITII Protein
A cDNA sequence encoding the CKMBITII was constructed and used to recombinantly synthesize the protein.
Marke: enQuireBio™ QP10554-1mg
Additional Details : Gewicht : 0.01000kg
Spezifikation
CKMBITII Protein | |
Research Use Only | |
The enzyme activity measured by kinetic assay at 340nm was 883 IU/mg at 37 degrees Celsius, corresponding to a Specific Activity of 1,133ng/ml. | |
Human | |
Untagged |
1 mg | |
< 1.0 EU per ug protein as determined by the LAL method. | |
Recombinant Protein | |
P. pastoris | |
Each mg of protein contains 20mM Tris-HCl, pH 6.8, 1mM EDTA and 1mM DTT, 50% (v/v) glycerol. |