missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human CKMBITII Protein
A cDNA sequence encoding the CKMBITII was constructed and used to recombinantly synthesize the protein.
131.00 CHF - 1735.00 CHF
Spezifikation
Name | CKMBITII Protein |
---|---|
Kennzeichnung | Research Use Only |
Endotoxin-Konzentration | < 1.0 EU per ug protein as determined by the LAL method. |
Biologische Aktivität | The enzyme activity measured by kinetic assay at 340nm was 883 IU/mg at 37 degrees Celsius, corresponding to a Specific Activity of 1,133ng/ml. |
Produkttyp | Recombinant Protein |
Produktcode | Marke | Menge | Preis | Menge & Verfügbarkeit | |||||
---|---|---|---|---|---|---|---|---|---|
Produktcode | Marke | Menge | Preis | Menge & Verfügbarkeit | |||||
15964228
|
enQuireBio™
QP10554-10UG |
10 μg |
131.00 CHF
10 Mikrogramm |
Verfügbar ab: 17-06-2024
Loggen Sie sich ein, um den Lagerbestand zu sehen |
|||||
15984228
|
enQuireBio™
QP10554-50UG |
50 μg |
203.00 CHF
50 Mikrogramm |
Verfügbar ab: 17-06-2024
Loggen Sie sich ein, um den Lagerbestand zu sehen |
|||||
15974228
|
enQuireBio™
QP10554-1MG |
1 mg |
1735.00 CHF
1 Milligramm |
Verfügbar ab: 17-06-2024
Loggen Sie sich ein, um den Lagerbestand zu sehen |
|||||
Spezifikation
CKMBITII Protein | |
< 1.0 EU per ug protein as determined by the LAL method. | |
Recombinant Protein | |
P. pastoris | |
Each mg of protein contains 20mM Tris-HCl, pH 6.8, 1mM EDTA and 1mM DTT, 50% (v/v) glycerol. |
Research Use Only | |
The enzyme activity measured by kinetic assay at 340nm was 883 IU/mg at 37 degrees Celsius, corresponding to a Specific Activity of 1,133ng/ml. | |
Human | |
Untagged |