missing translation for 'onlineSavingsMsg'
Learn More

Novus Biologicals™ Recombinant Human CD40 Ligand/TNFSF5 Protein

Product Code. 18642606 Shop All Bio Techne Products
missing translation for 'orderingAttributeHoverText'
Menge:
1 mg
10 μg
100 μg
500 μg
missing translation for 'unitSize'
1 Milligramm
10 Mikrogramm
100 Mikrogramm
500 Mikrogramm
This item is not returnable. View return policy

Product Code. 18642606

missing translation for 'mfr': Novus Biologicals™ NBP2352441mg

om dit product te kopen Registreer vandaag om een webaccount aan te maken

This item is not returnable. View return policy

Recombinant Protein

Fully biologically active when compared to standard. The ED50 as determined by the dose-dependent stimulation of IL-8 production by human PBMC is less than 5-10 ng/mL.
TRUSTED_SUSTAINABILITY

Specifications

Zugriffsnummer P29965, P29965, P29965
Zur Verwendung mit (Anwendung) SDS-PAGE, Bioactivity
Zusammensetzung Lyophilized from a 0.2 μm filtered concentrated solution in PBS, pH 7.0.
Gen-ID (Entrez) 959
Molekulargewicht MolecularWeight-theroretical: 16.3 kDa
Menge 1 mg
Lagerungsbedingungen Store at -20°C to -70°C as supplied. After reconstitution, store at 2 to 8 C for 1 month and (at -20°C to -70°C for long term storage. Avoid repeated freeze/thaw cycles.)
Endotoxin-Konzentration Less than 1 EU/μg of CD40 Ligand/TNFSF5 as determined by LAL method.
Gen-Alias CD154, CD154 antigen, CD40 antigen ligand, CD40 ligand, CD40 Ligand/TNFSF5, CD40-L, CD40LIGM, gp39, hCD40L, HIGM1, T-B cell-activating molecule, T-BAM, T-cell antigen Gp39, TNF-related activation protein, TNFSF5IMD3, TRAPtumor necrosis factor (ligand) superfamily, member 5 (hyper-IgM syndrome), tumor necrosis factor (ligand) superfamily member 5, Tumor necrosis factor ligand superfamily member 5
Gensymbol CD40LG
Rekonstitution Recommended to centrifuge prior to opening. Reconstitute in sterile distilled water or aqueous buffer containing 0.1% BSA to a concentration of 0.1-0.3 mg/mL Apportion stock solutions into working aliquots and store (at <-20°C.)
Forschungskategorie Biologically Active Proteins
Reinheits- oder Qualitätsgrad >95%, by SDS-PAGE and HPLC
Protein CD40 Ligand/TNFSF5
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.