missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human CD23 / FCER2 / FCER2A Protein
A DNA sequence encoding the Human CD23 / FCER2 / FCER2A, was expressed in the hosts and tags indicated.
Marke: enQuireBio™ QP10184-ec-20ug
Additional Details : Gewicht : 0.01000kg
Specifications
NP_001193948.2 | |
CD23 / FCER2 / FCER2A Protein | |
Research Use Only | |
FCER2 | |
The ED50 determined by its ability to induce TNF-alpha production by human PBMCs. | |
Human | |
Untagged | |
0.2 um filtered 20 mM PBS, pH 7.4, 150mM NaCl, lyophilized |
2208 | |
20 μg | |
Less than 1.0 EU/ug as determined by LAL method. | |
Partial (see sequence information for more details). | |
Recombinant Protein | |
E. coli | |
MELQVSSGFV CNTCPEKWIN FQRKCYYFGK GTKQWVHARY ACDDMEGQLV SIHSPEEQDF LTKHASHTGS WIGLRNLDLK GEFIWVDGSH VDYSNWAPGE PTSRSQGEDC VMMRGSGRWN DAFCDRKLGA WVCDRLATCT PPASEGSAES MGPDSRPDPD GRLPTPSAPL HS | |
Greater than 95% as determined by SDS-PAGE and HPLC. |