missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human CCL4 / MIP1B Protein

A DNA sequence encoding the Human CCL4 / MIP1B, was expressed in the hosts and tags indicated.

Marke:  enQuireBio™ QP10227-ec-250ug

Additional Details : Gewicht : 0.01000kg

 Lernen Sie weitere Varianten dieses

Artikelnummer. 15983148

  • 2310.00 CHF / 250 Mikrogramm


Verfügbar ab
30-10-2020
Zum Warenkorb hinzufügen

Spezifikation

Spezifikation

NP_002975.1
CCL4 / MIP1B Protein
250 μg
Less than 1.0 EU/ug as determined by LAL method.
Partial (see sequence information for more details).
Recombinant Protein
E. coli
APMGSDPPTA CCFSYTARKL PRNFVVDYYE TSSLCSQPAV VFQTKRGKQV CADPSESWVQ EYVYDLELN
388372
Greater than 97% as determined by SDS-PAGE and HPLC.
Research Use Only
CCL4
The ED50 as determined by a cell proliferation assay using human CCR5 transfected murine BaF3 cells is less than 2.0 ng/ml, corresponding to a specific activity of > 5.0 M 105 IU/mg.
Human
Untagged
0.2 um filtered PBS, pH 7.4, lyophilized
Videos
Dokumentation