missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human bFGF / FGF2 Protein
Recombinant Protein
Marke: enQuireBio™ QP5269-500ug
Additional Details : Gewicht : 0.01000kg
Spezifikation
NP_001997.5 | |
bFGF / FGF2 Protein | |
Research Use Only | |
FGF2 | |
Recombinant Protein | |
E. coli | |
MAAGSITTLP ALPEDGGSGA FPPGHFKDPK RLYCKNGGFF LRIHPDGRVD GVREKSDPHI KLQLQAEERG VVSIKGVCAN RYLAMKEDGR LLASKCVTDE CFFFERLESN NYNTYRSRKY TSWYVALKRT GQYKLGSKTG PGQKAILFLP MSAKS | |
Greater than 95% as determined by SDS-PAGE. |
2247 | |
500 μg | |
Less than or equal to 1.00 EUs/ug as determined by the LAL method. | |
Shows bioactivity at a concentration of 5 ng/mL in the 3T3 cell proliferation assay. The bioactivity acceptance criteria is set at 2 x 10^5 units/mg. | |
Human | |
Untagged | |
10 mM sodium phosphate, 75 mM sodium chloride, pH 7.5 |