missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human ARFIP1 341 a. a. Protein
A cDNA sequence encoding the ARFIP1 341 a. a. was constructed and used to recombinantly synthesize the protein.
Marke: enQuireBio™ QP11063-2ug
Additional Details : Gewicht : 0.01000kg
Spezifikation
27236 | |
2 μg | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Recombinant Protein | |
E. coli | |
MGSSHHHHHH SSGLVPRGSH MGSMAQESPK NSAAEIPVTS NGEVDDSREH SFNRDLKHSL PSGLGLSETQ ITSHGFDNTK EGVIEAGAFQ GGQRTQTKSG PVILADEIKN PAMEKLELVR KWSLNTYKCT RQIISEKLGR GSRTVDLELE AQIDILRDNK KKYENILKLA QTLSTQLFQM VHTQRQLGDA FADLSLKSLE LHEEFGYNAD TQKLLAKNGE TLLGAINFFI ASVNTLVNKT IEDTLMTVKQ YESARIEYDA YRTDLEELNL GPRDANTLPK IEQSQHLFQA HKEKYDKMRN DVSVKLKFLE ENKVKVLHNQ LVLFHNAIAA YFAGNQKQLE QTLKQFHIKL KTPGVDAPSW LEEQ | |
Greater than 90.0% as determined by SDS-PAGE. |
ARFIP1 341 a. a. Protein | |
Research Use Only | |
ARFIP1 | |
Human | |
His | |
ARFIP1 protein solution (0.25 mg/ml) contains Phosphate buffered saline (pH 7.4), 30% glycerol and 1mM DTT. |