missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human ANXA7 Protein

A cDNA sequence encoding the ANXA7 was constructed and used to recombinantly synthesize the protein.

Marke:  enQuireBio™ QP11036-5ug

Additional Details : Gewicht : 0.01000kg

 Lernen Sie weitere Varianten dieses

Artikelnummer. 15975938

  • 131.00 CHF / 5 Mikrogramm
Verfügbar ab: 01-07-2024
Sie sich ein, um den Lagerbestand zu sehen
Zum Warenkorb hinzufügen

Beschreibung

Beschreibung

Spezifikation

Spezifikation

ANXA7 Protein
Research Use Only
Recombinant Protein
E. coli
MGSSHHHHHH SSGLVPRGSH MGSHMSYPGY PPTGYPPFPG YPPAGQESSF PPSGQYPYPS GFPPMGGGAY PQVPSSGYPG AGGYPAPGGY PAPGGYPGAP QPGGAPSYPG VPPGQGFGVP PGGAGFSGYP QPPSQSYGGG PAQVPLPGGF PGGQMPSQYP GGQPTYPSQP ATVTQVTQGT IRPAANFDAI RDAEILRKAM KGFGTDEQAI VDVVANRSND QRQKIKAAFK TSYGKDLIKD LKSELSGNME ELILALFMPP TYYDAWSLRK AMQGAGTQER VLIEILCTRT NQEIREIVRC YQSEFGRDLE KDIRSDTSGH FERLLVSMCQ GNRDENQSIN HQMAQEDAQR LYQAGEGRLG TDESCFNMIL ATRSFPQLRA TMEAYSRMAN RDLLSSVSRE FSGYVESGLK TILQCALNRP AFFAERLYYA MKGAGTDDST LVRIVVTRSE IDLVQIKQMF AQMYQKTLGT MIAGDTSGDY RRLLLAIVGQ
Greater than 85.0% as determined by SDS-PAGE.
5 μg
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request.
Human
His
ANXA7 protein solution (0.25 mg/ml) containing 20mM Tris-HCl buffer (pH 8.0), 150mM NaCl, 1mM DTT and 40% glycerol.
Produktvorschläge

Produktvorschläge

Videos
Sicherheitsdatenblatt (SDS)
Dokumentation

Dokumentation

Zertifikate
Sonderangebote

Sonderangebote