missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human ANAPC13 Protein
A cDNA sequence encoding the ANAPC13 was constructed and used to recombinantly synthesize the protein.
Marke: enQuireBio™ QP11017-1mg
Additional Details : Gewicht : 0.01000kg
Spezifikation
ANAPC13 Protein | |
Research Use Only | |
Recombinant Protein | |
E. coli | |
MASMTGGQQM GRGSHMDSEV QRDGRILDLI DDAWREDKLP YEDVAIPLNE LPEPEQDNGG TTESVKEQEM KWTDLALQYL HENVPPIGN | |
Greater than 90.0% as determined by SDS-PAGE. |
1 mg | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Human | |
Untagged | |
The ANAPC13 solution (1 mg/ml) contains 20mM Tris-HCl buffer (pH 8.0), 20% glycerol, 1mM DTT and 0.1M NaCl. |