missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human ALDH6A1 Protein

A cDNA sequence encoding the ALDH6A1 was constructed and used to recombinantly synthesize the protein.

Marke:  enQuireBio™ QP11001-2ug

Additional Details : Gewicht : 0.01000kg

 Lernen Sie weitere Varianten dieses

Artikelnummer. 15985828

  • 131.00 CHF / 2 Mikrogramm
Verfügbar ab: 27-06-2024
Sie sich ein, um den Lagerbestand zu sehen
Zum Warenkorb hinzufügen

Beschreibung

Beschreibung

Spezifikation

Spezifikation

4329
2 μg
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request.
Recombinant Protein
E. coli
MGSSHHHHHH SSGLVPRGSH MGSSSSVPTV KLFIGGKFVE SKSDKWIDIH NPATNEVIGR VPQATKAEMD AAIASCKRAF PAWADTSVLS RQQVLLRYQQ LIKENLKEIA KLITLEQGKT LADAEGDVFR GLQVVEHACS VTSLMMGETM PSITKDMDLY SYRLPLGVCA GIAPFNFPAM IPLWMFPMAM VCGNTFLMKP SERVPGATML LAKLLQDSGA PDGTLNIIHG QHEAVNFICD HPDIKAISFV GSNKAGEYIF ERGSRHGKRV QANMGAKNHG VVMPDANKEN TLNQLVGAAF GAAGQRCMAL STAVLVGEAK KWLPELVEHA KNLRVNAGDQ PGADLGPLIT PQAKERVCNL IDSGTKEGAS ILLDGRKIKV KGYENGNFVG PTIISNVKPN MTCYKEEIFG PVLVVLETET LDEAIQIVNN NPYGNGTAIF TTNGATARKY AHLVDVGQVG VNVPIPVPLP MFSFTGSRSS FRGDTNFYGK QGIQFYTQLK TITSQWKEED ATLSSPAVVM PTMGR
Greater than 85.0% as determined by SDS-PAGE.
ALDH6A1 Protein
Research Use Only
ALDH6A1
Human
His
ALDH6A1 protein solution (0.25 mg/ml) containing Phosphate buffered saline (pH 7.4), 10% glycerol and 1mM DTT.
Produktvorschläge

Produktvorschläge

Videos
Sicherheitsdatenblatt (SDS)
Dokumentation

Dokumentation

Zertifikate
Sonderangebote

Sonderangebote