missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human ALDH6A1 Protein
A cDNA sequence encoding the ALDH6A1 was constructed and used to recombinantly synthesize the protein.
131.00 CHF - 1735.00 CHF
Spezifikation
Gen-ID (Entrez) | 4329 |
---|---|
Name | ALDH6A1 Protein |
Kennzeichnung | Research Use Only |
Endotoxin-Konzentration | Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. |
Gensymbol | ALDH6A1 |
Produktcode | Marke | Menge | Preis | Menge & Verfügbarkeit | |||||
---|---|---|---|---|---|---|---|---|---|
Produktcode | Marke | Menge | Preis | Menge & Verfügbarkeit | |||||
15985828
|
enQuireBio™
QP11001-2UG |
2 μg |
131.00 CHF
2 Mikrogramm |
Verfügbar ab: 17-06-2024
Loggen Sie sich ein, um den Lagerbestand zu sehen |
|||||
15975828
|
enQuireBio™
QP11001-10UG |
10 μg |
203.00 CHF
10 Mikrogramm |
Verfügbar ab: 17-06-2024
Loggen Sie sich ein, um den Lagerbestand zu sehen |
|||||
15965828
|
enQuireBio™
QP11001-100UG |
100 μg |
1735.00 CHF
100 Mikrogramm |
Verfügbar ab: 17-06-2024
Loggen Sie sich ein, um den Lagerbestand zu sehen |
|||||
Spezifikation
4329 | |
Research Use Only | |
ALDH6A1 | |
Human | |
His | |
ALDH6A1 protein solution (0.25 mg/ml) containing Phosphate buffered saline (pH 7.4), 10% glycerol and 1mM DTT. |
ALDH6A1 Protein | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Recombinant Protein | |
E. coli | |
MGSSHHHHHH SSGLVPRGSH MGSSSSVPTV KLFIGGKFVE SKSDKWIDIH NPATNEVIGR VPQATKAEMD AAIASCKRAF PAWADTSVLS RQQVLLRYQQ LIKENLKEIA KLITLEQGKT LADAEGDVFR GLQVVEHACS VTSLMMGETM PSITKDMDLY SYRLPLGVCA GIAPFNFPAM IPLWMFPMAM VCGNTFLMKP SERVPGATML LAKLLQDSGA PDGTLNIIHG QHEAVNFICD HPDIKAISFV GSNKAGEYIF ERGSRHGKRV QANMGAKNHG VVMPDANKEN TLNQLVGAAF GAAGQRCMAL STAVLVGEAK KWLPELVEHA KNLRVNAGDQ PGADLGPLIT PQAKERVCNL IDSGTKEGAS ILLDGRKIKV KGYENGNFVG PTIISNVKPN MTCYKEEIFG PVLVVLETET LDEAIQIVNN NPYGNGTAIF TTNGATARKY AHLVDVGQVG VNVPIPVPLP MFSFTGSRSS FRGDTNFYGK QGIQFYTQLK TITSQWKEED ATLSSPAVVM PTMGR | |
Greater than 85.0% as determined by SDS-PAGE. |