missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human ADAT2 Protein
A cDNA sequence encoding the ADAT2 was constructed and used to recombinantly synthesize the protein.
Marke: enQuireBio™ QP10964-2ug
Additional Details : Gewicht : 0.01000kg
Spezifikation
134637 | |
2 μg | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Recombinant Protein | |
E. coli | |
MGSSHHHHHH SSGLVPRGSH MEAKAAPKPA ASGACSVSAE ETEKWMEEAM HMAKEALENT EVPVGCLMVY NNEVVGKGRN EVNQTKNATR HAEMVAIDQV LDWCRQSGKS PSEVFEHTVL YVTVEPCIMC AAALRLMKIP LVVYGCQNER FGGCGSVLNI ASADLPNTGR PFQCIPGYRA EEAVEMLKTF YKQENPNAPK SKVRKKECQK S | |
Greater than 90% as determined by Analysis by SDS-PAGE. |
ADAT2 Protein | |
Research Use Only | |
ADAT2 | |
Human | |
His | |
The ADAT2 (0.5 mg/ml) protein contains 20mM Tris-HCl buffer pH 8.0, 2mM DTT, 50mM NaCl and 10% glycerol. |