missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human ADAM10 Protein
A cDNA sequence encoding the ADAM10 was constructed and used to recombinantly synthesize the protein.
Marke: enQuireBio™ QP10961-HIS-5ug
Additional Details : Gewicht : 0.01000kg
Spezifikation
ADAM10 Protein | |
Research Use Only | |
Recombinant Protein | |
E. coli | |
MVLLRVLILL LSWAAGMGGQ YGNPLNKYIR HYEGLSYNVD SLHQKHQRAK RAVSHEDQFL RLDFHAHGRH FNLRMKRDTS LFSDEFKVET SNKVLDYDTS HIYTGHIYGE EGSFSHGSVI DGRFEGFIQT RGGTFYVEPA ERYIKDRTLP FHSVIYHEDD INYPHKYGPQ GGCADHSVFE RMRKYQMTGV EEVTQIPQEE HAANGPELLR KKRTTSAEKN TCQLYIQTDH LFFKYYGTRE AVIAQISSHV KAIDTIYQTT DFSGIRNISF MVKRIRINTT ADEKDPTNPF RFPNIGVEKF LELNSEQNHD DYCLAYVFTD RDFDDGVLGL AWVGAPSGSS GGICEKSKLY SDGKKKSLNT GIITVQNYGS HVPPKVSHIT FAHEVGHNFG SPHDSGTECT PGESKNLGQK ENGNYIMYAR ATSGDKLNNN KFSLCSIRNI SQVLEKKRNN CFVESGQPIC GNGMVEQGEE CDCGYSDQCK DECCFDANQP EGRKCKLKPG KQCSPSQGPC CTAQCAFKSK SEKCRDDSDC AREGICNGFT ALCPASDPKP NFTDCNRHTQ VCINGQCAGS ICEKYGLEEC TCASSDGKDD KELCHVCCMK KMDPSTCAST GSVQWSRHFS GRTITLQPGS PCNDFRGYCD VFMRCRLVDA DGPLARLKKA IFSPELYENI AEWIVAHWWA VLLMGIALIM LMAGFIKICS VHTPSSNPKL PPPKPLPGTL KRRRPPQPIQ QPQRQRPRES YQMGHMRR | |
Greater than 95.0% as determined by SDS-PAGE. |
5 μg | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Human | |
His | |
The ADAM10 solution contains 20mM Tris (pH 8), 1mM EDTA and 50% Glycerol. |