missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human ACP6 Protein
A cDNA sequence encoding the ACP6 was constructed and used to recombinantly synthesize the protein.
Marke: enQuireBio™ QP10503-1mg
Additional Details : Gewicht : 0.01000kg
Spezifikation
ACP6 Protein | |
Research Use Only | |
Specific activity is > 70 units/mg, and is defined as the amount of enzyme which hydrolyze 1.0 nmoles of p-nitrophenyl phosphate (pNPP) per minute at PH 5.0 at 37C. | |
Human | |
His | |
ACP6 protein solution (1 mg/ml) containing Phosphate buffered saline (pH 7.4), 30% glycerol and 1mM DTT. |
1 mg | |
< 1.0 EU per ug protein as determined by the LAL method. | |
Recombinant Protein | |
E. coli | |
MGSSHHHHHH SSGLVPRGSH MGSELQEADG QCPVDRSLLK LKMVQVVFRH GARSPLKPLP LEEQVEWNPQ LLEVPPQTQF DYTVTNLAGG PKPYSPYDSQ YHETTLKGGM FAGQLTKVGM QQMFALGERL RKNYVEDIPF LSPTFNPQEV FIRSTNIFRN LESTRCLLAG LFQCQKEGPI IIHTDEADSE VLYPNYQSCW SLRQRTRGRR QTASLQPGIS EDLKKVKDRM GIDSSDKVDF FILLDNVAAE QAHNLPSCPM LKRFARMIEQ RAVDTSLYIL PKEDRESLQM AVGPFLHILE SNLLKAMDSA TAPDKIRKLY LYAAHDVTFI PLLMTLGIFD HKWPPFAVDL TMELYQHLES KEWFVQLYYH GKEQVPRGCP DGLCPLDMFL NAMSVYTLSP EKYHALCSQT QVMEVGNEE | |
Greater than 95.0% as determined by SDS-PAGE. |