missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant HSV HSV-2 gD Protein
A cDNA sequence encoding the HSV-2 gD was constructed and used to recombinantly synthesize the protein.
Marke: enQuireBio™ QP12344-100ug
Weitere Details : Gewicht : 0.01000kg
Spezifikation
HSV HSV-2 gD Protein | |
Research Use Only | |
Recombinant Protein | |
E. coli | |
MANUAL | |
Protein is >95% pure as determined by SDS-PAGE |
100 μg | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
HSV | |
His | |
50% glycerol, 1.5 M Urea, 0.1% SDS and 100 mM NaCl. |