Learn More
enQuireBio™ Recombinant Dengue Virus Serotype 3 VLP Protein
Recombinant Protein
Marke:  enQuireBio™ QP10481-100ug
Additional Details : Gewicht : 0.01000kg
 Lernen Sie weitere Varianten dieses
Artikelnummer. 15973998
  • 1240.00 CHF / 100 Mikrogramm
Verfügbar ab: 25-06-2024
Sie sich ein, um den Lagerbestand zu sehen
Zum Warenkorb hinzufügen
Beschreibung
Beschreibung
This VLP protein is combinantion of the C, E1 and E2 sequences. Virus-Like Particles (VLPs) are a new form of protein expression that consist of protein shells with outer proteins that are specific to the given virus. VLPs are immunogenic but are non-infectious, lacking the primary genetic material of the virus. Additionally, VLPs more effectively activate aspects of the immune response, which improves immune stimulation and immunological memory. VLP-based vaccines have also shown to provide effective protection and are in current use for several diseases.
Spezifikation
Spezifikation
Serotype 3 VLP Protein
Research Use Only
Recombinant Protein
Human
10mM sodium phosphate, 20mM sodium citrate, 154mM sodium chloride, pH7.4.
100 μg
Untested
Dengue virus (Strain: Sri Lanka D3/H/IMTSSA-SRI/2000/1266)
Native
>95% as determined by SDS-PAGE.