missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Bovine Trypsin Protein
A cDNA sequence encoding the Trypsin was constructed and used to recombinantly synthesize the protein.
Marca: enQuireBio™ QP10896-1mg
Dettagli aggiuntivi : Gewicht : 0.01000kg
Specifica
Trypsin Protein | |
Research Use Only | |
3,650 Units/mg. | |
Bovine | |
Untagged | |
Greater than 90% as determined by:(a) Analysis by RP-HPLC. (b) Analysis by SDS-PAGE. |
1 mg | |
< 1.0 EU per ug protein as determined by the LAL method. | |
Recombinant Protein | |
Bovine | |
The protein was lyophilized without any additives. |