missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Bovine GH Protein
A cDNA sequence encoding the GH was constructed and used to recombinantly synthesize the protein.
Marke: enQuireBio™ QP10639-20ug
Additional Details : Gewicht : 0.01000kg
Spezifikation
GH Protein | |
Research Use Only | |
The activity as determined by the PDFP13B9 cells stably transfected with rabbit GH receptors. Bovine GH is also capable of forming a 1:2 complex with the recombinant ovine growth hormone receptor extracellular domain (ECD). | |
Bovine | |
Untagged | |
The Bovine Growth Hormone protein was lyophilized from a concentrated (1 mg/ml) solution with 0.0045mM NaHCO3 adjusted to pH 8-9. |
20 μg | |
< 1.0 EU per ug protein as determined by the LAL method. | |
Recombinant Protein | |
E. coli | |
1 afpamslsgl fanavlraqh lhqlaadtfk efertyipeg qrysiqntqv 51 afcfsetmpa ptgkneaqqk sdlellrisl lliqswlgpl qflsrvftns 101 lvfgtsdrvy eklkdleegi lalmreledg tprrgqilkq tydkfdtnmr 151 sddallknyg llscfrkdlh ktetylrvmk crrfgeasca f | |
Greater than 98.0% as determined by:(a) Analysis by SEC-HPLC. (b) Analysis by SDS-PAGE. |