Learn More
Rabbit anti-Goat IgG (H+L) Secondary Antibody, Novus Biologicals™
Artikelnummer. 18002424 Alle ansehen Bio Techne Produkte
Change view
Click to view available options
Menge:
2 mg
Packungsgröße:
2 Milligramm
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Menge unitSize
18002424 2 mg 2 Milligramm
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Artikelnummer. 18002424 Lieferant Novus Biologicals Lieferanten-Nr. NBP172707
om dit product te kopen Registreer vandaag om een webaccount aan te maken
Rabbit Polyclonal Antibody
IgG (H+L) Polyclonal specifically detects IgG (H+L) in Goat samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Fluorophore-linked immunosorbent assay.
TRUSTED_SUSTAINABILITY
Spezifikation
Antigen IgG (H+L)
Anwendungen Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry, Immunofluorescence
Klassifikation Polyclonal
Konjugat Unconjugated
Verdünnung Western Blot 1:10000 - 1:60000, ELISA 1:800000, Immunohistochemistry 1:1000 - 1:5000, Immunocytochemistry/Immunofluorescence, Fluorophore-linked immunosorbent assay
Wirtsspezies Rabbit
Immunogen Goat IgG whole molecule
Reinigungsverfahren Affinity Purified
Menge 2 mg
Regulatorischer Status RUO
Forschungsgebiet Adaptive Immunity, Immunology
Primär oder sekundär Secondary
Testspezifität This product was prepared from monospecific antiserum by immunoaffinity chromatography using Goat IgG coupled to agarose beads. Assay by immunoelectrophoresis resulted in a single precipitin arc against anti-Rabbit Serum, Goat IgG and Goat Serum.
Zielspezies Goat
Inhalt und Lagerung Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Isotype IgG