Learn More
IKA R 402 Stirring Shaft Protection
Artikelnummer. 31910313 Alle ansehen IKA Produkte
Ansicht ändern
Click to view available options
Material:
Plastic
Packungsgröße:
1 Stück
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Material Packungsgröße
31910313 Plastic 1 Stück
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Artikelnummer. 31910313 Lieferant IKA Lieferanten-Nr. 0020115312
  • ONLINE EXCLUSIVE PRICE
    163.09 CHF
    153.07 CHF / 1 Stück
    Sparen 10.02 CHF
    6% Rabatt
Lager vorrätig
Zum Warenkorb hinzufügen
Zum Warenkorb hinzufügen
missing translation for 'validMainframeMsg'
Stirring shaft protection for overhead stirrers to prevent users from potential injuries at the rotating shafts and stirring elements.
  • Equipped with a hinge to facilitate access to the stirrer chuck without having to remove it from the motor housing.
  • Can be attached directly to the overhead stirrers RW 20 digital, RW 28 digital, and the devices of the EUROSTAR series.
TRUSTED_SUSTAINABILITY
Spezifikation
Typ Chuck Guard
Material Plastic