Learn More
PTMS Rabbit anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marke: Proteintech 12106-1-AP-150UL
Additional Details : Gewicht : 0.00010kg
Beschreibung
Product No. 12106-1-AP is a rabbit polyclonal antibody raised against the full-length of human PTMS. The MW of this protein is 12 kDa, and this antibody specifically recognizes the 12 kDa protein.
PTMS may mediate immune function by blocking the effect of prothymosin alpha which confers resistance to certain opportunistic infections.Spezifikation
PTMS | |
Polyclonal | |
Unconjugated | |
PTMS | |
ParaT, parathymosin, PTMS | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
5763 | |
-20°C | |
Liquid |
Immunocytochemistry, Immunofluorescence, Western Blot | |
0.13 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
P20962 | |
PTMS | |
PTMS Fusion Protein Ag2751 | |
150 μL | |
Primary | |
Human | |
Antibody | |
IgG |