missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PTMS Rabbit anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
133.80 CHF - 381.34 CHF
Spezifikation
Antigen | PTMS |
---|---|
Konzentration | 0.13 mg/mL |
Anwendungen | Immunocytochemistry, Western Blot, Immunofluorescence |
Klassifikation | Polyclonal |
Konjugat | Unconjugated |
Produktcode | Marke | Menge | Preis | Menge & Verfügbarkeit | |||||
---|---|---|---|---|---|---|---|---|---|
Produktcode | Marke | Menge | Preis | Menge & Verfügbarkeit | |||||
16839623
|
Proteintech
12106-1-AP-20UL |
20 μL |
133.80 CHF
20 Mikroliter |
Verfügbar ab: 07-05-2024
Loggen Sie sich ein, um den Lagerbestand zu sehen |
|||||
16829623
|
Proteintech
12106-1-AP-150UL |
150 μL |
381.34 CHF
150 Mikroliter |
Verfügbar ab: 07-05-2024
Loggen Sie sich ein, um den Lagerbestand zu sehen |
|||||
Beschreibung
Product No. 12106-1-AP is a rabbit polyclonal antibody raised against the full-length of human PTMS. The MW of this protein is 12 kDa, and this antibody specifically recognizes the 12 kDa protein.
PTMS may mediate immune function by blocking the effect of prothymosin alpha which confers resistance to certain opportunistic infections.Spezifikation
PTMS | |
Immunocytochemistry, Western Blot, Immunofluorescence | |
Unconjugated | |
Rabbit | |
Human | |
P20962 | |
5763 | |
PTMS Fusion Protein Ag2751 | |
Primary | |
-20°C | |
PTMS |
0.13 mg/mL | |
Polyclonal | |
Liquid | |
RUO | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
ParaT, parathymosin, PTMS | |
PTMS | |
IgG | |
Antigen Affinity Chromatography | |
Antibody |