missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Abnova™ PSMD8 (Human) Recombinant Protein
Human PSMD8 full-length ORF ( NP_002803, 1 a.a. - 257 a.a.) recombinant protein with GST-tag at N-terminal.
Marke: Abnova™ H00005714-P01.10ug
Additional Details : Gewicht : 0.00010kg
Beschreibung
- Sequence: MYEQLKGEWNRKSPNLSKCGEELGRLKLVLLELNFLPTTGTKLTKQQLILARDILEIGAQWSILRKDIPSFERYMAQLKCYYFDYKEQLPESAYMHQLLGLNLLFLLSQNRVAEFHTELERLPAKDIQTNVYIKHPVSLEQYLMEGSYNKVFLAKGNIPAESYTFFIDILLDTIRDEIAGCIEKAYEKILFTEATRILFFNTPKKMTDYAKKRGWVLGPNNYYSFASQQQKPEDTTIPSTELAKQVIEYARQLEMIV
Spezifikation
NP_002803 | |
proteasome (prosome, macropain) 26S subunit, non-ATPase, 8 | |
125% SDS-PAGE Stained with Coomassie Blue | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
PSMD8 | |
GST |
5714 | |
Wheat germ expression system | |
10 μg | |
HIP6, HYPF, MGC1660, Nin1p, Rpn12, S14, p31 | |
Wheat Germ (in vitro) | |
50 mM Tris-HCI, 10 mM reduced Glutathione, pH 8.0 in the elution buffer |