missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PSMD12 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marke: Proteintech 11412-1-AP-20UL
Additional Details : Gewicht : 0.00010kg
Beschreibung
This protein is a ubiquitously expressed nuclear protein and belongs to a highly conserved subfamily of WD-repeat proteins. It is found among several proteins that binds directly to retinoblastoma protein, which regulates cell proliferation. The encoded protein is found in many histone deacetylase complexes, including mSin3 co-repressor complex. It is also present in protein complexes involved in chromatin assembly. This protein can interact with BRCA1 tumor-suppressor gene and may have a role in the regulation of cell proliferation and differentiation.Spezifikation
PSMD12 | |
Polyclonal | |
Unconjugated | |
PSMD12 | |
p55, PSMD12, Rpn5 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
287772, 5718, 66997 | |
-20°C | |
Liquid |
Immunocytochemistry, Immunohistochemistry (Paraffin), Immunofluorescence, Western Blot | |
0.13 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
O00232, Q9D8W5 | |
Psmd12 | |
PSMD12 Fusion Protein Ag1957 | |
20 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |