Learn More
PPP3R1 Rabbit anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marke: Proteintech 13210-1-AP-150UL
Dieser Artikel kann nicht zurückgegeben werden.
Rückgaberichtlinie anzeigen
Beschreibung
Protein phosphatase 3 (PPP3, formerly PP2B, Calcineurin), a serine/ threonine protein phosphatase, is a heterodimer composed of one catalytic subunit (PPP3C, Calcineurin A) and one regulatory subunit (PPP3R, Calcineurin B). PPP3R has two isoforms, PPP3R1 and PPP3R2. PPP3Rl is ubiquitously expressed in different tissues and is coded by the PPP3R1 gene which was mapped recently to human chromosome 2. PPP3Rl has four functional calcium-binding sites.Spezifikation
| PPP3R1 | |
| Polyclonal | |
| Unconjugated | |
| PPP3R1 | |
| Calcineurin subunit B type 1, CALNB1, CNA2, CNB, CNB1, PPP3R1 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 5534 | |
| -20°C | |
| Liquid |
| Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot | |
| 0.23 mg/mL | |
| PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
| P63098 | |
| PPP3R1 | |
| PPP3R1 Fusion Protein Ag3992 | |
| 150 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |