missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PPP3CB Mouse anti-Human, Mouse, Porcine, Rat, Clone: 3C1A3, Proteintech
Mouse Monoclonal Antibody
Marke: Proteintech 66614-1-IG-150UL
Additional Details : Gewicht : 0.00010kg
Beschreibung
PPP3CB is a calcium-dependent, calmodulin-stimulated protein phosphatase. This subunit may have a role in the calmodulin activation of calcineurin.Spezifikation
PPP3CB | |
Monoclonal | |
1 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
P16298, P20651, P48453 | |
Ppp3cb | |
PPP3CB Fusion Protein Ag26987 | |
150 μL | |
Primary | |
Human, Mouse, Porcine, Rat | |
Antibody | |
IgG1 |
Western Blot | |
3C1A3 | |
Unconjugated | |
PPP3CB | |
CALNA2, CALNB, CAM PRP catalytic subunit, CNA2, PPP3CB | |
Mouse | |
Protein G | |
RUO | |
19056, 24675, 396603, 5532 | |
-20°C | |
Liquid |