Learn More
Semadeni™ Einwegspritzen aus Polypropylen
Mit Luer-Lock-Adaptern.
Marke: Semadeni™ 1961
Additional Details : Gewicht : 0.01000kg
Beschreibung
- Three-piece design
- With black silicone gasket
- Available in 5, 10 and 20 mL volumes
- Manufactured from polypropylene (PP)
Spezifikation
Three-piece | |
Yes | |
20 mL | |
Yes |
Black silicone gasket | |
50/Pk. | |
Luer |