missing translation for 'onlineSavingsMsg'
Learn More
Learn More
POLR2A Rabbit anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marke: Proteintech 20655-1-AP-20UL
Additional Details : Gewicht : 0.00010kg
Beschreibung
This gene encodes the largest subunit of RNA polymerase II, the polymerase responsible for synthesizing messenger RNA in eukaryotes. The product of this gene contains a carboxy terminal domain composed of heptapeptide repeats that are essential for polymerase activity. These repeats contain serine and threonine residues that are phosphorylated in actively transcribing RNA polymerase. In addition, this subunit, in combination with several other polymerase subunits, forms the DNA binding domain of the polymerase, a groove in which the DNA template is transcribed into RNA.Spezifikation
POLR2A | |
Polyclonal | |
Unconjugated | |
POLR2A | |
hRPB220, hsRPB1, Pol II, POLR2, POLR2A, POLRA, RNA polymerase II subunit B1, RPB1, RPBh1, RpIILS, RPO2, RPOL2 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
5430 | |
-20°C | |
Liquid |
Western Blot | |
0.33 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
P24928 | |
POLR2A | |
Peptide | |
20 μL | |
Primary | |
Human | |
Antibody | |
IgG |