missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PMM1 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marke: Proteintech 11252-2-AP-20UL
Additional Details : Gewicht : 0.00010kg
Beschreibung
PMM1, also named as PMMH22, belongs to the eukaryotic PMM family. It catalyzes the second step in the conversion of fructose-6P to GDP-mannose is and is involved in the synthesis of the GDP-mannose and dolichol-phosphate-mannose required for a number of critical mannosyl transfer reactions. In addition, PMM1 may be responsible for the degradation of glucose-1, 6-bisphosphate in ischemic brain.Spezifikation
PMM1 | |
Polyclonal | |
Unconjugated | |
PMM1 | |
phosphomannomutase 1, PMM 1, PMM1, PMMH 22, PMMH22, Sec53 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
29858, 300089, 5372 | |
-20°C | |
Liquid |
Western Blot | |
0.17 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
O35621, Q92871 | |
PMM1 | |
PMM1 Fusion Protein Ag1782 | |
20 μL | |
Primary | |
Human, Rat, Mouse | |
Antibody | |
IgG |