missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PKIG Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marke: Proteintech 21371-1-AP-20UL
Additional Details : Gewicht : 0.00010kg
Beschreibung
Extremely potent competitive inhibitor of cAMP-dependent protein kinase activity, this protein interacts with the catalytic subunit of the enzyme after the cAMP-induced dissociation of its regulatory chains.Spezifikation
PKIG | |
Polyclonal | |
Unconjugated | |
PKIG | |
PKI gamma, PKIG | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
11142, 18769, 266709 | |
-20°C | |
Liquid |
Western Blot, Immunofluorescence, Immunocytochemistry | |
0.25 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
O70139, Q9Y2B9 | |
Pkig | |
PKIG Fusion Protein Ag15878 | |
20 μL | |
Primary | |
Human, Rat, Mouse | |
Antibody | |
IgG |