missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ PI4KA Human Protein
PI4KB and PI4KA are the two human type III PI4 kinases.
Marke: Invitrogen™ PV5690
Additional Details : Gewicht : 0.01000kg
Beschreibung
PI4KB and PI4KA share sequence similarly with PI3 kinases and phosphorylate the 4 position of phosphotidyl inositol.Spezifikation
NP_002641.1 | |
50mM Tris pH 7.5, 150mM NaCl, 0.02% Trition X-100, 0.5mM EDTA, 2mM DTT, 50% Glycerol. | |
1mg | |
RUO - research use only | |
PI4KA | |
Human | |
Baculovirus |
Enzyme and Protein Activity Assay, Kinase Assay, Protein Assays and Analysis, Protein Biology | |
124.4kDa | |
Store at -80°C. At first use, aliquot and store at -80°C to avoid multiple freeze-thaws | |
Full Length | |
PI3 Kinase Family | |
GST Tag | |
Frozen |