missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ PI4KA Human Protein
PI4KB and PI4KA are the two human type III PI4 kinases.
Marke: Invitrogen™ PV5689
Additional Details : Gewicht : 0.00010kg
Beschreibung
PI4KB and PI4KA share sequence similarly with PI3 kinases and phosphorylate the 4 position of phosphotidyl inositol.Spezifikation
NP_002641.1 | |
50mM Tris pH 7.5, 150mM NaCl, 0.02% Trition X-100, 0.5mM EDTA, 2mM DTT, 50% Glycerol. | |
124.0kDa | |
Store at -80°C. At first use, aliquot and store at -80°C to avoid multiple freeze-thaws | |
Full Length | |
PI4KA, phosphatidylinositol 4-kinase, catalytic, alpha | |
Human | |
Baculovirus | |
Frozen |
Enzyme and Protein Activity Assay, Industrial and Applied Science, Kinase Assay, Kinase Biology, Kinase Proteins and Expression Reagents, Pharma and Biopharma, Protein Assays and Analysis, Protein Biology, Target and Lead Identification and Validation | |
PI4KA | |
20μg | |
N-Terminal | |
PI4KA | |
PI3 Kinase Family | |
GST Tag | |
Kinases & Inhibitors | |
Lipid Kinases |