Learn More
Phospho-p70(S6K) (Thr229) Rabbit anti-Human, Unconjugated, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marke: Proteintech 28988-1-AP-20UL
Beschreibung
The Phospho-p70(S6K) (Thr229) antibody from Proteintech is a rabbit polyclonal antibody to a peptide of human Phospho-p70(S6K) (Thr229). This antibody recognizes human antigen. The Phospho-p70(S6K) (Thr229) antibody has been validated for the following applications: WB, ELISA analysis.
Spezifikation
| Phospho-p70(S6K) (Thr229) | |
| Polyclonal | |
| Rabbit | |
| Primary | |
| Human |
| Western Blot, ELISA | |
| Unconjugated | |
| 20 μL | |
| 6198 | |
| IgG |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.