missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PHF19 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marke: Proteintech 11895-1-AP-150UL
Additional Details : Gewicht : 0.00010kg
Beschreibung
PHF19 contains 2 PHD-type zinc fingers. It acts as a transcritpional repressor. Isoform 1 and isoform 2 inhibit transcription from an HSV-tk promoter.Spezifikation
PHF19 | |
Polyclonal | |
Unconjugated | |
PHF19 | |
hPCL3, MTF2L1, PCL3, PHD finger protein 19, PHF19, Polycomb like protein 3 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
26147, 296653, 74016 | |
-20°C | |
Liquid |
Immunocytochemistry, Immunofluorescence, Western Blot | |
0.44 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
Q5T6S3, Q9CXG9 | |
Phf19 | |
PHF19 Fusion Protein Ag2511 | |
150 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |