missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PDCD6IP Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
133.80 CHF - 381.34 CHF
Spezifikation
Antigen | PDCD6IP |
---|---|
Konzentration | 0.14 mg/mL |
Anwendungen | Immunocytochemistry, Immunofluorescence, Western Blot, Immunoprecipitation |
Klassifikation | Polyclonal |
Konjugat | Unconjugated |
Produktcode | Marke | Menge | Preis | Menge & Verfügbarkeit | |||||
---|---|---|---|---|---|---|---|---|---|
Produktcode | Marke | Menge | Preis | Menge & Verfügbarkeit | |||||
16850134
|
Proteintech
12422-1-AP-20UL |
20 μL |
133.80 CHF
20 Mikroliter |
Verfügbar ab: 14-05-2024
Loggen Sie sich ein, um den Lagerbestand zu sehen |
|||||
16840134
|
Proteintech
12422-1-AP-150UL |
150 μL |
381.34 CHF
150 Mikroliter |
Verfügbar ab: 14-05-2024
Loggen Sie sich ein, um den Lagerbestand zu sehen |
|||||
Beschreibung
Apoptotic linked-gene-product 2 (ALG-2) interacting protein X (ALIX) is a conserved adaptor protein which is ubiquitously expressed. Alix was originally reported to play a role in apoptosis but has recently been shown to be involved in other cellular mechanisms including endosomal sorting, endocytosis, viral buding and actin cytoskeleton assembly.Spezifikation
PDCD6IP | |
Immunocytochemistry, Immunofluorescence, Western Blot, Immunoprecipitation | |
Unconjugated | |
Rabbit | |
Mouse, Rat, Human | |
Q8WUM4, Q9QZA2, Q9WU78 | |
10015, 18571, 501083 | |
PDCD6IP Fusion Protein Ag3074 | |
Primary | |
-20°C | |
PDCD6IP |
0.14 mg/mL | |
Polyclonal | |
Liquid | |
RUO | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
AIP1, ALG 2 interacting protein 1, Alix, DRIP4, HP95, KIAA1375, PDCD6 interacting protein, PDCD6IP | |
Pdcd6ip | |
IgG | |
Antigen Affinity Chromatography | |
Antibody |