missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PDCD6IP Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marke: Proteintech 12422-1-AP-20UL
Additional Details : Gewicht : 0.00010kg
Beschreibung
Apoptotic linked-gene-product 2 (ALG-2) interacting protein X (ALIX) is a conserved adaptor protein which is ubiquitously expressed. Alix was originally reported to play a role in apoptosis but has recently been shown to be involved in other cellular mechanisms including endosomal sorting, endocytosis, viral buding and actin cytoskeleton assembly.Spezifikation
PDCD6IP | |
Polyclonal | |
Unconjugated | |
PDCD6IP | |
AIP1, ALG 2 interacting protein 1, Alix, DRIP4, HP95, KIAA1375, PDCD6 interacting protein, PDCD6IP | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
10015, 18571, 501083 | |
-20°C | |
Liquid |
Immunocytochemistry, Immunofluorescence, Western Blot, Immunoprecipitation | |
0.14 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
Q8WUM4, Q9QZA2, Q9WU78 | |
Pdcd6ip | |
PDCD6IP Fusion Protein Ag3074 | |
20 μL | |
Primary | |
Mouse, Rat, Human | |
Antibody | |
IgG |