missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PDCD6 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marke: Proteintech 12303-1-AP-150UL
Additional Details : Gewicht : 0.00010kg
Beschreibung
An increased intracellular Ca2+ concentration induces apoptotic cell death. Transiently elevated Ca2+ concentrations are required for glucocorticoid-mediated and T cell receptor-mediated pathways, leading to T cell apoptosis. ALG-2 (for apoptosis-linked gene 2) is a Ca2+ -binding protein that participates in regulatory events occuring late in the apoptotic program and where several death signals converge. ALG-2 is a protein expressed in normal brain, and to a greater extent in ischemic brain. The ALG-2 protein contains five EF-hand-like motifs and shares homology with members of the penta EF-hand family, which includes Calpain small subunits sorcin and Grancalcin.Spezifikation
PDCD6 | |
Polyclonal | |
Unconjugated | |
PDCD6 | |
ALG 2, ALG2, PDCD6, PEF1B, programmed cell death 6 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
10016, 18570, 308061 | |
-20°C | |
Liquid |
Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot | |
0.37 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
G3V7W1, O75340, P12815 | |
Pdcd6 | |
PDCD6 Fusion Protein Ag2949 | |
150 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |