Learn More
Panendothelial Cell Antigen Antibody (MECA-32), Alexa Fluor™ 488, Novus Biologicals™
Artikelnummer. 18711510 Alle ansehen Bio Techne Produkte
Change view
Click to view available options
Menge:
0.1 mL
0.5 mL
Packungsgröße:
0.10 Milliliter
0.50 Milliliter
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Menge unitSize
18711510 0.1 mL 0.10 Milliliter
18722853 0.5 mL 0.50 Milliliter
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Artikelnummer. 18711510 Lieferant Novus Biologicals Lieferanten-Nr. NB10077668AF488SS
om dit product te kopen Registreer vandaag om een webaccount aan te maken
Rat Monoclonal Antibody
Panendothelial Cell Antigen Monoclonal antibody specifically detects Panendothelial Cell Antigen in Human, Mouse samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence, Immunoprecipitation, Immunohistochemistry (Paraffin), Immunohistochemistry (Frozen), CyTOF
TRUSTED_SUSTAINABILITY
Spezifikation
Antigen Panendothelial Cell Antigen
Anwendungen Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunoprecipitation
Klassifikation Monoclonal
Klon MECA-32
Konjugat Alexa Fluor 488
Zusammensetzung 50mM Sodium Borate
Gen-Alias FELS, Fenestrated endothelial-linked structure protein, gp68, MECA32, MECA-32, Panendothelial Cell Antigen, plasmalemma vesicle associated protein, Plasmalemma Vesicle Protein 1, PV-1 protein
Wirtsspezies Rat
Immunogen Mouse lymph node stromal cells.
Reinigungsverfahren Protein G purified
Menge 0.1 mL
Regulatorischer Status RUO
Forschungsgebiet Cancer, Cell Biology, Cellular Markers, Endothelial Cell Markers, Extracellular Matrix, Hypoxia, Immunology, Mesenchymal Stem Cell Markers, Stem Cell Markers
Primär oder sekundär Primary
Gen-ID (Entrez) 84094
Zielspezies Human, Mouse
Inhalt und Lagerung Store at 4°C in the dark.
Form Purified
Isotype IgG2a κ