missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PAMCI Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marke: Proteintech 12861-1-AP-20UL
Additional Details : Gewicht : 0.00010kg
Beschreibung
The protein encoded by this gene localizes to perinuclear endosomes. This protein associates with peptidylglycine alpha-amidating monooxygenase, and may be involved with the trafficking of this enzyme through secretory or endosomal pathways.Spezifikation
PAMCI | |
Polyclonal | |
Unconjugated | |
RASSF9 | |
P CIP1, PAMCI, PCIP1, RASSF9 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
237504, 65053, 9182 | |
-20°C | |
Liquid |
Western Blot, Immunocytochemistry, Immunofluorescence | |
0.27 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
O75901, O88869, Q8K342 | |
Rassf9 | |
PAMCI Fusion Protein Ag3976 | |
20 μL | |
Primary | |
Rat, Human, Mouse | |
Antibody | |
IgG |