Learn More
Novus Biologicals™ PACSIN3 Overexpression Lysate
Artikelnummer. 18296073 Alle ansehen Bio Techne Produkte
Ansicht ändern
Click to view available options
Menge:
0,1 mg
Packungsgröße:
0.10 Milligramm
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Menge unitSize
18296073 0,1 mg 0.10 Milligramm
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Artikelnummer. 18296073 Lieferant Novus Biologicals™ Lieferanten-Nr. NBL114060
Please to purchase this item. Need a web account? Register with us today!
The PACSIN3 Overexpression Lysate is derived from human. It has been validated for the following applications: Western Blot.
TRUSTED_SUSTAINABILITY
Spezifikation
Zelllinie The lysate was created in HEK293T cells, using plasmid ID RC2413 and based on accession number NM_16223. The protein contains a C-terminal DDK tag.
Zur Verwendung mit (Anwendung) Western Blot
Menge 0,1 mg
Lagerungspuffer RIPA buffer
Reinheit Protein
Testspezifität Homo sapiens protein kinase C and casein kinase substrate in neurons 3 (PACSIN3), mRNA.
Inhalt und Lagerung -80°C. Avoid freeze-thaw cycles.
Protein State Native
Nur für Forschungszwecke