missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Osteoglycin Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marke: Proteintech 12755-1-AP-20UL
Additional Details : Gewicht : 0.00010kg
Beschreibung
Mimecan induces ectopic bone formation in conjunction with transforming growth factor beta. Mimecan is a small proteoglycan which contains tandem leucine-rich repeats. Mimecan is encoded by the OGN gene, a member of the small leucine-rich proteoglycan (SLRP) family of proteins. Mimecan induces ectopic bone formation in conjunction with transforming growth factor beta (TGF-beta-1 or TGF-beta-2) and may regulate osteoblast differentiation. High expression of mimecan protein may be associated with elevated heart left ventricular mass. Alternative splicing results in multiple transcript variants of mimecan. Alternatively spliced transcript variants that encode mimecan have been found.Spezifikation
Osteoglycin | |
Polyclonal | |
Unconjugated | |
OGN | |
Mimecan, OG, OGN, OIF, Osteoglycin, Osteoinductive factor, SLRR3A | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
18295, 291015, 4969 | |
-20°C | |
Liquid |
Western Blot, Immunohistochemistry (Paraffin), Immunocytochemistry, Immunofluorescence | |
0.14 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
P20774, Q62000 | |
Ogn | |
Osteoglycin Fusion Protein Ag3484 | |
20 μL | |
Primary | |
Mouse, Rat, Human | |
Antibody | |
IgG |