missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Osteocalcin Rabbit anti-Mouse, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marke: Proteintech 20277-1-AP-150UL
Additional Details : Gewicht : 0.00010kg
Beschreibung
The antibody is specific to mouse Bglap1.
Osteocalcin belongs to the osteocalcin/matrix Gla-protein family and constitutes 1-2% of the total bone protein. It binds strongly to apatite and calcium. Gamma-carboxyglutamate residues are formed by vitamin K dependent carboxylation. These residues are essential for the binding of calcium.Spezifikation
Osteocalcin | |
Polyclonal | |
Unconjugated | |
Bglap | |
Bglap, Bglap1, mOC A, OC, OG1, Osteocalcin | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
12096 | |
-20°C | |
Liquid |
Immunohistochemistry (Paraffin) | |
0.17 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
P86546 | |
Bglap | |
Peptide | |
150 μL | |
Primary | |
Mouse | |
Antibody | |
IgG |