missing translation for 'onlineSavingsMsg'
Learn More
Learn More
OSBPL9 Rabbit anti-Human, Mouse, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marke: Proteintech 11879-1-AP-20UL
Additional Details : Gewicht : 0.00010kg
Beschreibung
This gene encodes a member of a subfamily of LIM domain proteins that are characterized by an N-terminal proline rich region and three C-terminal LIM domains. The encoded protein localizes to the cell periphery in focal adhesions and may be involved in cell-cell adhesion and cell motility. This protein also shuttles through the nucleus and may function as a transcriptional co-activator. This gene is located at the junction of certain disease related chromosomal translocations which result in the expression of chimeric proteins that may promote tumor growth. Alternate splicing results in multiple transcript variants.Spezifikation
OSBPL9 | |
Polyclonal | |
Unconjugated | |
OSBPL9 | |
ORP 9, ORP9, OSBP related protein 9, OSBP4, OSBPL9 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
100273, 114883 | |
-20°C | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot | |
0.13 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
A2A8Z1, Q96SU4 | |
OSBPL9 | |
OSBPL9 Fusion Protein Ag2476 | |
20 μL | |
Primary | |
Human, Mouse | |
Antibody | |
IgG |