missing translation for 'onlineSavingsMsg'
Learn More
Learn More
OGT Mouse anti-Human, Mouse, Porcine, Rat, Clone: 2B2A6, Proteintech
Mouse Monoclonal Antibody
Marke: Proteintech 66823-1-IG-20UL
Additional Details : Gewicht : 0.00010kg
Beschreibung
O-linked N-acetylglucosamine (O-GlcNAc), is a member of the O-GlcNAc transferase family. O-GlcNAc is a posttranslational modification characterized by the attachment of N-acetylglucosamine to specific serine/threonine hydroxyl groups. It plays an important role in nutrient sensing, gene expression, and protein degradation.Spezifikation
OGT | |
Monoclonal | |
1 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
O15294, P56558, Q27HV0 | |
OGT | |
OGT Fusion Protein Ag28402 | |
20 μL | |
Primary | |
Porcine, Human, Rat, Mouse | |
Antibody | |
IgG2a |
Western Blot | |
2B2A6 | |
Unconjugated | |
OGT | |
FLJ23071, HRNT1, O GLCNAC, OGT | |
Mouse | |
Protein A | |
RUO | |
108155, 26295, 664652, 8473 | |
-20°C | |
Liquid |