missing translation for 'onlineSavingsMsg'
Learn More
Learn More
OGFOD2 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marke: Proteintech 21590-1-AP-150UL
Additional Details : Gewicht : 0.00010kg
Beschreibung
OGFOD2 is a member of the 2-OG-Fe(II) dioxygenase family which plays an important role in ischemic survival and stress. Like the related protein OGFOD1, OGFOD2 is an oxidoreductase containing a Prolyl 4-hydroxylase alpha subunit. Little is known about the function of OGFOD2, but its similarity to OGFOD1 suggests that it is a novel stress granule component that may play an important proapoptotic role in the regulation of translation and HRI-mediated phosphorylation of eIF2alpha in cells.Spezifikation
OGFOD2 | |
Polyclonal | |
Unconjugated | |
OGFOD2 | |
OGFOD2 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
288657, 66627, 79676 | |
Antibody | |
IgG |
Immunohistochemistry (Paraffin), Western Blot | |
0.29 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
Q6N063 | |
Ogfod2 | |
OGFOD2 Fusion Protein Ag16164 | |
150 μL | |
Primary | |
Human, Mouse, Rat | |
Liquid |