missing translation for 'onlineSavingsMsg'
Learn More

NY-ESO-1 Rabbit anti-Human, Polyclonal, Proteintech

Rabbit Polyclonal Antibody

Marke:  Proteintech 19521-1-AP-20UL

Additional Details : Gewicht : 0.00010kg

 Lernen Sie weitere Varianten dieses

Artikelnummer. 16839054

  • 133.80 CHF / 20 Mikroliter
Verfügbar ab: 17-05-2024
Sie sich ein, um den Lagerbestand zu sehen
Zum Warenkorb hinzufügen

Beschreibung

Beschreibung

NY-ESO-1, a member of the CT (cancer/testis) family, is a tumor specific shared antigen with distinctive immunogenicity. The molecular basis of T cell-mediated antitumor immunity has been elucidated by the identification of a number of tumor antigens recognized by CD8+ T cells. Among these antigens, NY-ESO-1 is of particular interest because both the cytotoxic T lymphocyte and antibody have been shown to react with this antigen. The NY-ESO-1 has been detected in normal testis and in a range of human tumor types. NY-ESO-1 reactivity in testis is restricted germ cells, particularly in the spermatogonia.
Spezifikation

Spezifikation

NY-ESO-1
Polyclonal
Unconjugated
CTAG1A
Cancer/testis antigen 1, cancer/testis antigen 1A, Cancer/testis antigen 6.1, CT6.1, CTAG, CTAG1, CTAG1A, CTAG1B, ESO1, L antigen family member 2, LAGE 2, LAGE2, LAGE2A, LAGE2B, NY-ESO-1
Rabbit
Antigen Affinity Chromatography
RUO
246100
-20°C
Liquid
Western Blot, Immunofluorescence, Immunocytochemistry, Immunoprecipitation
0.24 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
P78358
CTAG1A
Peptide
20 μL
Primary
Human
Antibody
IgG
Produktvorschläge

Produktvorschläge

Videos
Sicherheitsdatenblatt (SDS)
Dokumentation

Dokumentation

Zertifikate
Sonderangebote

Sonderangebote