missing translation for 'onlineSavingsMsg'
Learn More
Learn More
NY-ESO-1 Rabbit anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
133.80 CHF - 381.34 CHF
Spezifikation
Antigen | NY-ESO-1 |
---|---|
Konzentration | 0.24 mg/mL |
Anwendungen | Western Blot, Immunofluorescence, Immunocytochemistry, Immunoprecipitation |
Klassifikation | Polyclonal |
Konjugat | Unconjugated |
Produktcode | Marke | Menge | Preis | Menge & Verfügbarkeit | |||||
---|---|---|---|---|---|---|---|---|---|
Produktcode | Marke | Menge | Preis | Menge & Verfügbarkeit | |||||
16839054
|
Proteintech
19521-1-AP-20UL |
20 μL |
133.80 CHF
20 Mikroliter |
Verfügbar ab: 13-05-2024
Loggen Sie sich ein, um den Lagerbestand zu sehen |
|||||
16829054
|
Proteintech
19521-1-AP-150UL |
150 μL |
381.34 CHF
150 Mikroliter |
Verfügbar ab: 13-05-2024
Loggen Sie sich ein, um den Lagerbestand zu sehen |
|||||
Beschreibung
NY-ESO-1, a member of the CT (cancer/testis) family, is a tumor specific shared antigen with distinctive immunogenicity. The molecular basis of T cell-mediated antitumor immunity has been elucidated by the identification of a number of tumor antigens recognized by CD8+ T cells. Among these antigens, NY-ESO-1 is of particular interest because both the cytotoxic T lymphocyte and antibody have been shown to react with this antigen. The NY-ESO-1 has been detected in normal testis and in a range of human tumor types. NY-ESO-1 reactivity in testis is restricted germ cells, particularly in the spermatogonia.Spezifikation
NY-ESO-1 | |
Western Blot, Immunofluorescence, Immunocytochemistry, Immunoprecipitation | |
Unconjugated | |
Rabbit | |
Human | |
P78358 | |
246100 | |
Peptide | |
Primary | |
-20°C | |
CTAG1A |
0.24 mg/mL | |
Polyclonal | |
Liquid | |
RUO | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
Cancer/testis antigen 1, cancer/testis antigen 1A, Cancer/testis antigen 6.1, CT6.1, CTAG, CTAG1, CTAG1A, CTAG1B, ESO1, L antigen family member 2, LAGE 2, LAGE2, LAGE2A, LAGE2B, NY-ESO-1 | |
CTAG1A | |
IgG | |
Antigen Affinity Chromatography | |
Antibody |